missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyanoadamantane 98.0+%, TCI America™
$82.79 - $302.25
Chemical Identifiers
| CAS | 23074-42-2 |
|---|---|
| Molecular Formula | C11H15N |
| Molecular Weight (g/mol) | 161.248 |
| MDL Number | MFCD00074731 |
| InChI Key | FQFZASRJFRAEIH-UHFFFAOYSA-N |
| Synonym | 1-adamantanecarbonitrile, 1-cyanoadamantane, 1-adamantyl cyanide, tricyclo 3.3.1.13,7 decane-1-carbonitrile, tricyclo 3.3.1.1∼3,7∼ decane-1-carbonitrile, adamantanecarbonitrile, 3r,5s,7s-adamantane-1-carbonitrile, 1-adamantylnitrile, pubchem8757 |
| PubChem CID | 90878 |
| IUPAC Name | adamantane-1-carbonitrile |
| SMILES | C1C2CC3CC1CC(C2)(C3)C#N |
Chemical Identifiers
| 23074-42-2 | |
| 161.248 | |
| FQFZASRJFRAEIH-UHFFFAOYSA-N | |
| 90878 | |
| C1C2CC3CC1CC(C2)(C3)C#N |
| C11H15N | |
| MFCD00074731 | |
| 1-adamantanecarbonitrile, 1-cyanoadamantane, 1-adamantyl cyanide, tricyclo 3.3.1.13,7 decane-1-carbonitrile, tricyclo 3.3.1.1∼3,7∼ decane-1-carbonitrile, adamantanecarbonitrile, 3r,5s,7s-adamantane-1-carbonitrile, 1-adamantylnitrile, pubchem8757 | |
| adamantane-1-carbonitrile |
Specifications
| 23074-42-2 | |
| White-Yellow | |
| MFCD00074731 | |
| 3439 | |
| FQFZASRJFRAEIH-UHFFFAOYSA-N | |
| adamantane-1-carbonitrile | |
| 90878 | |
| ≥98.0% (GC) | |
| 1-Cyanoadamantane |
| 195°C | |
| C11H15N | |
| 1 g | |
| 1-adamantanecarbonitrile, 1-cyanoadamantane, 1-adamantyl cyanide, tricyclo 3.3.1.13,7 decane-1-carbonitrile, tricyclo 3.3.1.1∼3,7∼ decane-1-carbonitrile, adamantanecarbonitrile, 3r,5s,7s-adamantane-1-carbonitrile, 1-adamantylnitrile, pubchem8757 | |
| C1C2CC3CC1CC(C2)(C3)C#N | |
| 161.248 | |
| 161.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : No