missing translation for 'onlineSavingsMsg'
Learn More

1-Cyanoadamantane 98.0+%, TCI America™

Catalog No. C07831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07831G 1 g
C07835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07831G Supplier TCI America Supplier No. C07831G

Chemical Identifiers

CAS 23074-42-2
Molecular Formula C11H15N
Molecular Weight (g/mol) 161.248
MDL Number MFCD00074731
InChI Key FQFZASRJFRAEIH-UHFFFAOYSA-N
Synonym 1-adamantanecarbonitrile, 1-cyanoadamantane, 1-adamantyl cyanide, tricyclo 3.3.1.13,7 decane-1-carbonitrile, tricyclo 3.3.1.1∼3,7∼ decane-1-carbonitrile, adamantanecarbonitrile, 3r,5s,7s-adamantane-1-carbonitrile, 1-adamantylnitrile, pubchem8757
PubChem CID 90878
IUPAC Name adamantane-1-carbonitrile
SMILES C1C2CC3CC1CC(C2)(C3)C#N

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 161.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Cyanoadamantane