missing translation for 'onlineSavingsMsg'
Learn More

1-Cyano-4-(dimethylamino)pyridinium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. C2348100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2348100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2348100MG Supplier TCI America Supplier No. C2348100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used for the electrophilic cyanation of Grignard reagents.

Chemical Identifiers

CAS 59016-56-7
Molecular Formula C8H10BF4N3
Molecular Weight (g/mol) 234.99
MDL Number MFCD00011998
InChI Key MBLVMDCQDCVKNE-UHFFFAOYSA-N
Synonym cdap, 1-cyano-4-dimethylamino pyridinium tetrafluoroborate, 1-cyano-4-dimethylamino pyridin-1-ium tetrafluoroborate, unii-p4w72066jt, 1-cyano-4-dimethylamino-pyridinium tetrafluoroborate, pubchem17735, acmc-209m9a, n-cyano-4-dimethylaminopyridinium tetrafluoroborate, 1-cyano-4-dimethylaminopyridinium tetra-fluoroborate, n-cyano-4-dimethylamino-pyridinium tetrafluoroborate
PubChem CID 9881151
IUPAC Name 1-cyano-4-(dimethylamino)pyridin-1-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN(C)C1=CC=[N+](C=C1)C#N

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 100 mg
UN Number 1759
Formula Weight 234.99
Percent Purity ≥98.0% (N)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 1-Cyano-4-(dimethylamino)pyridinium Tetrafluoroborate
TSCA No
Recommended Storage Freezer