missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorophthalazine, 97%
Description
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.
Solubility
Insoluble in water. Soluble in dichloromethane, ethanol and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Water, heat, moisture sensitive.
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.
Solubility
Insoluble in water. Soluble in dichloromethane, ethanol and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Water, heat, moisture sensitive.
Specifications
Specifications
| Melting Point | 109°C to 112°C |
| Quantity | 1 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Insoluble in water. Soluble in dichloromethane,ethanol and methanol. |
| Formula Weight | 164.59 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-Chlorophthalazine |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only