missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorophthalazine, 97%

Catalog No. AAH2714303
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2714303 1 g
AAH2714309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2714303 Supplier Thermo Scientific Chemicals Supplier No. H2714303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5784-45-2 | C8H5ClN2 | 164.592 g/mol

1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.

Solubility
Insoluble in water. Soluble in dichloromethane, ethanol and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Water, heat, moisture sensitive.

Chemical Identifiers

CAS 5784-45-2
Molecular Formula C8H5ClN2
Molecular Weight (g/mol) 164.592
MDL Number MFCD00024141
InChI Key UCOVESIAFFGEOR-UHFFFAOYSA-N
Synonym phthalazine, 1-chloro, 1-chloro-phthalazine, chlorophthalazine, ccris 7361, 1-chloro-2,3-benzodiazine, 1chlorophthalazine, 1-chlorophthalazin, acmc-20a2sh, 1-chloro phthalazine, 1-chlorophthalazine 1g
PubChem CID 160793
IUPAC Name 1-chlorophthalazine
SMILES C1=CC=C2C(=C1)C=NN=C2Cl

Specifications

Melting Point 109°C to 112°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Insoluble in water. Soluble in dichloromethane,ethanol and methanol.
Formula Weight 164.59
Percent Purity 97%
Chemical Name or Material 1-Chlorophthalazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only