missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropentane 99.0+%, TCI America™

Catalog No. p-7124302
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C023725ML 25 mL
C0237500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C023725ML Supplier TCI America Supplier No. C023725ML
Only null left

Chemical Identifiers

CAS 543-59-9
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.59
MDL Number MFCD00001015
InChI Key SQCZQTSHSZLZIQ-UHFFFAOYSA-N
Synonym pentyl chloride, pentane, 1-chloro, n-amyl chloride, n-pentyl chloride, amyl chloride, chloropentane, unii-0eg9msd3nk, 0eg9msd3nk, pentane, chloro, n-butylcarbonyl chloride
PubChem CID 10977
IUPAC Name 1-chloropentane
SMILES CCCCCCl

Specifications

Color Colorless
Boiling Point 108°C
Quantity 25 mL
UN Number 1107
Formula Weight 106.59
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloropentane