missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroisoquinoline, 97+%

Catalog No. AC391380010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391380010 1 g Glass bottle
AC391380050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC391380010 Supplier Thermo Scientific Chemicals Supplier No. 391380010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19493-44-8 | C9H6ClN | 163.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19493-44-8
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.61
MDL Number MFCD00024134
InChI Key MSQCQINLJMEVNJ-UHFFFAOYSA-N
Synonym 1-chloro-isoquinoline, isoquinoline, 1-chloro, chloroisoquinoline, isoquinoline, chloro, 1-chloroisoquinone, 1-chloro isoquinoline, zlchem 1129, pubchem13906, acmc-1bowu, 1-chloroisoquinoline
PubChem CID 140539
IUPAC Name 1-chloroisoquinoline
SMILES C1=CC=C2C(=C1)C=CN=C2Cl

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 31°C to 36°C
Color White to Yellow
Boiling Point 274°C to 275°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.61
Percent Purity 97+%
Physical Form Low Melting Solid, Crystals and/or Chunks
Chemical Name or Material 1-Chloroisoquinoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only