missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloroheptane 99.0+%, TCI America™
Supplier: TCI America C0182500ML
Specifications
| 1-Chloroheptane | |
| -69°C | |
| 159°C | |
| MFCD00001021 | |
| 1993 | |
| DZMDPHNGKBEVRE-UHFFFAOYSA-N | |
| 1-chloroheptane | |
| 12371 | |
| ≥99.0% (GC) |
| 629-06-1 | |
| Yellow | |
| C7H15Cl | |
| 500 mL | |
| Heptyl Chloride | |
| CCCCCCCCl | |
| 134.65 | |
| 134.65 | |
| Liquid |
Chemical Identifiers
| 629-06-1 | |
| 134.65 | |
| DZMDPHNGKBEVRE-UHFFFAOYSA-N | |
| 12371 | |
| CCCCCCCCl |
| C7H15Cl | |
| MFCD00001021 | |
| Heptyl Chloride | |
| 1-chloroheptane |
Safety and Handling
EINECSNumber : (2)-0066
TSCA : Yes