missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroheptane 99.0+%, TCI America™

Catalog No. C018225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C018225ML 25 mL
C0182500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C018225ML Supplier TCI America Supplier No. C018225ML
Only null left

Chemical Identifiers

CAS 629-06-1
Molecular Formula C7H15Cl
Molecular Weight (g/mol) 134.65
MDL Number MFCD00001021
InChI Key DZMDPHNGKBEVRE-UHFFFAOYSA-N
Synonym Heptyl Chloride
PubChem CID 12371
IUPAC Name 1-chloroheptane
SMILES CCCCCCCCl

Specifications

Melting Point -69°C
Color Yellow
Boiling Point 159°C
Quantity 25 mL
UN Number 1993
Formula Weight 134.65
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloroheptane
EINECSNumber (2)-0066
TSCA Yes