missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroethyl Ethyl Carbonate 98.0+%, TCI America™

Catalog No. C105625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C105625G 25 g
C1056100G 100 g
C1056500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C105625G Supplier TCI America Supplier No. C105625G

Chemical Identifiers

CAS 50893-36-2
Molecular Formula C5H9ClO3
Molecular Weight (g/mol) 152.574
MDL Number MFCD00040523
InChI Key YVRGKFXJZCTTRB-UHFFFAOYSA-N
Synonym 1-chlorodiethyl carbonate, carbonic acid, 1-chloroethyl ethyl ester, 1-chloroethyl ethylcarbonate, 1-ethoxycarbonyloxyethyl chloride, carbonic acid 1-chloroethyl ethyl ester, 1-chloroethylethylcarbonate, acmc-1aw9h, 1-chloroethyl ethoxyformate, ethyl 1-chloroethyl carbonate, ksc490c2n
PubChem CID 162095
IUPAC Name 1-chloroethyl ethyl carbonate
SMILES CCOC(=O)OC(C)Cl

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 g
Formula Weight 152.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloroethyl Ethyl Carbonate
EINECSNumber (2)-3303
TSCA Yes