missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorodocosane 98.0+%, TCI America™

Catalog No. C07755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07755G 5 g
C077525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07755G Supplier TCI America Supplier No. C07755G
Only null left

Chemical Identifiers

CAS 42217-03-8
Molecular Formula C22H45Cl
Molecular Weight (g/mol) 345.05
MDL Number MFCD00039383
InChI Key OACXFSZVCDOBKF-UHFFFAOYSA-N
Synonym Docosyl Chloride
PubChem CID 545602
IUPAC Name 1-chlorodocosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCl

Specifications

Melting Point 41°C
Color White-Yellow
Quantity 5 g
Formula Weight 345.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chlorodocosane
EINECSNumber (2)-0066&(2)-0068
TSCA No