missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorodecane 97.0+%, TCI America™

Catalog No. C0600500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0600500ML 500 mL
C060025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0600500ML Supplier TCI America Supplier No. C0600500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1002-69-3
Molecular Formula C10H21Cl
Molecular Weight (g/mol) 176.73
MDL Number MFCD00000956
InChI Key ZTEHOZMYMCEYRM-UHFFFAOYSA-N
Synonym decyl chloride, decane, 1-chloro, n-decyl chloride, decane, chloro, unii-l0hsc44e5x, decyl chloride mixed isomers, chlorodecane, l0hsc44e5x, chlorodecane mixed isomers, 1-chlorodecane
PubChem CID 13848
IUPAC Name 1-chlorodecane
SMILES CCCCCCCCCCCl

Specifications

Melting Point -34°C
Color Yellow
Boiling Point 223°C
Quantity 500 mL
UN Number 3082
Formula Weight 176.73
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorodecane
EINECSNumber (2)-0066&(2)-0068
RTECSNumber HD7050000
TSCA Yes