missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorodecane, 97-99%

Catalog No. AC108982500
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108982500 250 mL Glass bottle
AC108980010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108982500 Supplier Thermo Scientific Chemicals Supplier No. 108982500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1002-69-3 | C10H21Cl | 176.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1002-69-3
Molecular Formula C10H21Cl
Molecular Weight (g/mol) 176.73
MDL Number MFCD00000956
InChI Key ZTEHOZMYMCEYRM-UHFFFAOYSA-N
Synonym decyl chloride, decane, 1-chloro, n-decyl chloride, decane, chloro, unii-l0hsc44e5x, decyl chloride mixed isomers, chlorodecane, l0hsc44e5x, chlorodecane mixed isomers, 1-chlorodecane
PubChem CID 13848
IUPAC Name 1-chlorodecane
SMILES CCCCCCCCCCCl

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point -34°C
Color Colorless
Density 0.8600g/mL
Boiling Point 223°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4362 to 1.4382
Linear Formula CH3(CH2)9Cl
Quantity 250 mL
Beilstein 01, 168
Specific Gravity 0.86
Solubility Information Solubility in water: 0.01g/L (20°C)
Viscosity 1.94 mPa.s (20°C)
Formula Weight 176.73
Percent Purity 97-99%
Physical Form Liquid
Chemical Name or Material 1-Chlorodecane, 97-99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-691-0
TSCA TSCA

RUO – Research Use Only