missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane 99.0+%, TCI America™

Catalog No. C0142500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0142500ML 500 mL
C014225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0142500ML Supplier TCI America Supplier No. C0142500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Melting Point -123°C
Color Colorless
Boiling Point 78°C
Quantity 500 mL
UN Number 1127
Formula Weight 92.57
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorobutane
EINECSNumber (2)-0060
RTECSNumber EJ6300000
TSCA Yes