missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane, HPLC Grade, 99.5+%

Maximum level of impurities: Evaporation residue 1ppm, H2O 0.02% | CAS: 109-69-3 | C4H9Cl | 92.57 g/mol

Supplier:  Thermo Scientific Chemicals 039198K7

 View more versions of this product

Catalog No. AA39198K7


Only null left
Explore available promotions

Description

Description

1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as alkylating reagent in organic and organometallic compound synthesis. It is also employed as an antihelmintic in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Chlorobutane
109-69-3
Colorless
77°C to 78°C
280°C (536°F)
Pungent
1.4024
4 L
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
VFWCMGCRMGJXDK-UHFFFAOYSA-N
1-chlorobutane
8005
110 hPa (83mm Hg) at 20°C
1-Chlorobutane
Filtered through 0.2μ filters.
°C to 123°C
0.886 g/cm3 at 20°C
-12°C (10°F)
>99.5%
C4H9Cl
MFCD00001009
1730909
Miscible with alcohol and ether. Immiscible with water.
CCCCCl
92.57
92.57
HPLC
Liquid

Chemical Identifiers

109-69-3
92.57
VFWCMGCRMGJXDK-UHFFFAOYSA-N
8005
CCCCCl
C4H9Cl
MFCD00001009
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
1-chlorobutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H225-H304

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 203-696-6

RTECSNumber : EJ6300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only