missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane, HPLC Grade, 99.5+%

Catalog No. AA39198M1
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39198M1 4 x 1 L
AA39198AK 250 mL
AA39198K2 1 L
AA39198K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA39198M1 Supplier Thermo Scientific Chemicals Supplier No. 039198M1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Maximum level of impurities: Evaporation residue 1ppm, H2O 0.02% | CAS: 109-69-3 | C4H9Cl | 92.57 g/mol

1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as alkylating reagent in organic and organometallic compound synthesis. It is also employed as an antihelmintic in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Melting Point °C to 123°C
Color Colorless
Density 0.886
Boiling Point 77°C to 78°C
Flash Point -12°C (10°F)
Ignition Point 280°C (536°F)
Odor Pungent
Assay Percent Range >99.5%
Refractive Index 1.4024
Quantity 4 x 1 L
Beilstein 1730909
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Vapor Pressure 110 hPa (83mm Hg) at 20°C
Formula Weight 92.57
Grade HPLC
Concentration or Composition (by Analyte or Components) 1-Chlorobutane
Physical Form Liquid
Chemical Name or Material 1-Chlorobutane, Filtered through 0.2μ filters.
Show More Show Less
Hazard Category H225-H304
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-696-6
RTECSNumber EJ6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only