missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane, MilliporeSigma™

Catalog No. MCX09141
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MCX09141 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MCX09141 Supplier Technidata Supplier No. CX09141

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Density 0.886g/cm3 (20°C)
Explosion Limit 1.8 to 10.1%(V)
Ignition Point 280°C
Packaging Glass Bottle
Linear Formula CH3(CH2)3Cl
Quantity 4 L
Vapor Pressure 110 hPa (20°C)
Formula Weight 92.57
Chemical Name or Material 1-Chlorobutane, For Synthesis
Recommended Storage Store at +15°C to +25°C