missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane CHROMASOLV™, for HPLC, ≥99.8%, Solstice

Catalog No. 6000095
Change view
Click to view available options
Quantity:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000095 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000095 Supplier Solstice Supplier No. 349582L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™, for HPLC,≥99.8%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Melting Point -123°C
Density 0.886g/mL at 25°C (lit.)
Boiling Point 77°C to 78°C
Refractive Index n20/D 1.402
Linear Formula CH3(CH2)3Cl
Quantity 2 L
Beilstein 1730909
Vapor Pressure 80.1mmHg (78.4°C)
Formula Weight 92.57g/mol
Percent Purity ≥99.8%
Grade CHROMASOLV
Chemical Name or Material 1-Chlorobutane, for HPLC
Show More Show Less
Hazard Statement P210-P273-P301 + P310-P331-P403 + P235
Precautionary Statement H225-H304-H336-H412
EINECSNumber 203-696-6
RTECSNumber EJ6300000