missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane, 99.8%, for HPLC

Catalog No. AC425880010
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425880010 1 L Glass bottle
AC425880025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425880010 Supplier Thermo Scientific Chemicals Supplier No. 425880010
Get 1 Extra Chemical When You Buy 2
View all promotions

CAS: 109-69-3 | C4H9Cl | 92.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Acidity 0.0003 meq/g max.
Absorbance 1 cm cell vs HPLC grade water, 0.05 max. at 240nm, 0.01 max. at 250nm, 0.005 max. at 260nm, 0.005 max. at 270nm, 1 cm cell vs HPLC grade water at 0nm
CAS Min % 99.75
CAS Max % 100.0
Melting Point -123.0°C
Color Colorless
Density 0.8800g/mL
Boiling Point 77.0°C to 78.0°C
Flash Point −12°C
Assay Percent Range 99.7% min. (GC)
Packaging Glass bottle
Refractive Index 1.4014 to 1.4034
Linear Formula CH3(CH2)3Cl
Quantity 1 L
UN Number 1127
Residue after Evaporation 2ppm max.
Specific Gravity 0.88
Merck Index 15, 1561
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: miscible with alcohol and ether
Viscosity 0.45 mPa.s (20°C)
Formula Weight 92.57
Percent Purity 99.8%
Grade HPLC
Physical Form Liquid
Chemical Name or Material 1-Chlorobutane, For HPLC
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
EINECSNumber 203-696-6

RUO – Research Use Only