missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroanthraquinone 98.0+%, TCI America™

Catalog No. p-7124547
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C065325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C065325G Supplier TCI America Supplier No. C065325G
Only null left

Chemical Identifiers

CAS 82-44-0
Molecular Formula C14H7ClO2
Molecular Weight (g/mol) 242.658
MDL Number MFCD00001189
InChI Key BOCJQSFSGAZAPQ-UHFFFAOYSA-N
Synonym 1-chloroanthraquinone, 1-chloro anthraquinone, anthraquinone, 1-chloro, 1-chloro-9,10-anthraquinone, 9,10-anthracenedione, 1-chloro, 1-chloranthrachinon, 1-chloranthrachinon czech, alpha-chloroanthraquinone, alpha-monochloroanthraquinone, unii-96s5sqv15v
PubChem CID 6709
IUPAC Name 1-chloroanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Cl

Specifications

Melting Point 164°C
Quantity 25 g
Formula Weight 242.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloroanthraquinone
EINECSNumber (4)-0688
RTECSNumber CB6150000
TSCA Yes