missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloroanthraquinone, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 82-44-0 |
|---|---|
| Molecular Formula | C14H7ClO2 |
| Molecular Weight (g/mol) | 242.66 |
| MDL Number | MFCD00001189 |
| InChI Key | BOCJQSFSGAZAPQ-UHFFFAOYSA-N |
| Synonym | 1-chloroanthraquinone, 1-chloro anthraquinone, anthraquinone, 1-chloro, 1-chloro-9,10-anthraquinone, 9,10-anthracenedione, 1-chloro, 1-chloranthrachinon, 1-chloranthrachinon czech, alpha-chloroanthraquinone, alpha-monochloroanthraquinone, unii-96s5sqv15v |
| PubChem CID | 6709 |
| IUPAC Name | 1-chloroanthracene-9,10-dione |
| SMILES | C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC108600050
|
Thermo Scientific Chemicals
108600050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 82-44-0 | |
| 242.66 | |
| BOCJQSFSGAZAPQ-UHFFFAOYSA-N | |
| 6709 | |
| C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Cl |
| C14H7ClO2 | |
| MFCD00001189 | |
| 1-chloroanthraquinone, 1-chloro anthraquinone, anthraquinone, 1-chloro, 1-chloro-9,10-anthraquinone, 9,10-anthracenedione, 1-chloro, 1-chloranthrachinon, 1-chloranthrachinon czech, alpha-chloroanthraquinone, alpha-monochloroanthraquinone, unii-96s5sqv15v | |
| 1-chloroanthracene-9,10-dione |
Specifications
| 82-44-0 | |
| 100.0 | |
| 97.5% min. (GC) | |
| C14H7ClO2 | |
| 5g | |
| 1-chloroanthraquinone, 1-chloro anthraquinone, anthraquinone, 1-chloro, 1-chloro-9,10-anthraquinone, 9,10-anthracenedione, 1-chloro, 1-chloranthrachinon, 1-chloranthrachinon czech, alpha-chloroanthraquinone, alpha-monochloroanthraquinone, unii-96s5sqv15v | |
| BOCJQSFSGAZAPQ-UHFFFAOYSA-N | |
| 1-chloroanthracene-9,10-dione | |
| 6709 | |
| 98% |
| 97.5 | |
| Authentic | |
| Glass Bottle | |
| MFCD00001189 | |
| 07,787 | |
| Solubility in water: insoluble | |
| C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Cl | |
| 242.66 | |
| 242.66 | |
| 1-Chloroanthraquinone, 98% |
Safety and Handling
EINECSNumber : 201-421-4
RTECSNumber : CB6150000
TSCA : TSCA
RUO â Research Use Only