missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroadamantane 98.0+%, TCI America™

Catalog No. C07475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07475G 5 g
C074725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07475G Supplier TCI America Supplier No. C07475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 935-56-8
Molecular Formula C10H15Cl
Molecular Weight (g/mol) 170.68
MDL Number MFCD00075627
InChI Key OZNXTQSXSHODFR-UHFFFAOYSA-N
Synonym 1-adamantyl chloride, adamantyl chloride, 3-chloroadamantane, adamantane, 1-chloro, tricyclo 3.3.1.13,7 decane, 1-chloro, 1-chlorotricyclo 3.3.1.13,7 decane, 3r,5s,7s-1-chloroadamantane, 1-chlorotricyclo 3.3.1.1∼3,7∼ decane
PubChem CID 64154
IUPAC Name 1-chloroadamantane
SMILES ClC12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 166°C
Color White
Quantity 5 g
Formula Weight 170.68
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloroadamantane