missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroadamantane, 98%

Catalog No. p-7049581
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AA1516906 5 g
AA1516914 25 g
AA1516922 100 g
3 options

Catalog No. AA1516906

Supplier: Thermo Scientific Chemicals 01516906

Only null left

CAS: 935-56-8 | C10H15Cl | 170.68 g/mol

It is used in some dry etching masks and polymer formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 935-56-8
Molecular Formula C10H15Cl
Molecular Weight (g/mol) 170.68
MDL Number MFCD00075627
InChI Key OZNXTQSXSHODFR-UHFFFAOYSA-N
Synonym 1-adamantyl chloride, adamantyl chloride, 3-chloroadamantane, adamantane, 1-chloro, tricyclo 3.3.1.13,7 decane, 1-chloro, 1-chlorotricyclo 3.3.1.13,7 decane, 3r,5s,7s-1-chloroadamantane, 1-chlorotricyclo 3.3.1.1∼3,7∼ decane
PubChem CID 64154
IUPAC Name 1-chloroadamantane
SMILES ClC12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 165°C
Quantity 5 g
Solubility Information Poorly soluble in water. Soluble in hydrocarbons. But readily soluble in nonpolar organic solvents.
Formula Weight 170.68
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-Chloroadamantane
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-305-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only