missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-6-iodohexane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. p-7125613
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27145G 5 g
C271425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27145G Supplier TCI America Supplier No. C27145G
Only null left

Chemical Identifiers

CAS 34683-73-3
Molecular Formula C6H12ClI
Molecular Weight (g/mol) 246.516
MDL Number MFCD00039417
InChI Key QTJHNJCILMMRIQ-UHFFFAOYSA-N
Synonym hexane, 1-chloro-6-iodo, hexamethylene chloroiodide, 6-chlorohexyl iodide, acmc-20aovr, 6-chloro-1-iodohexane, 1-iodo-6-chloro hexane, 1-chloro-6-iodohexane, 1-chloro-6-iodohexane, stab. with copper chip, stabilized with copper chip
PubChem CID 118713
IUPAC Name 1-chloro-6-iodohexane
SMILES C(CCCI)CCCl

Specifications

Boiling Point 92°C
Quantity 5 g
Formula Weight 246.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-6-iodohexane (stabilized with Copper chip)