missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-6-hydroxyhexane, 95%

CAS: 2009-83-8 | C6H13ClO | 136.62 g/mol

Supplier:  Thermo Scientific Chemicals 109281000

 View more versions of this product

Catalog No. AC109281000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-chloro-6-hydroxyhexane
94.0
Colorless to Yellow
108.0°C to 112.0°C (14.0 mmHg)
Authentic
Glass bottle
1.4547 to 1.4567
MFCD00002984
1.03
JNTPTNNCGDAGEJ-UHFFFAOYSA-N
6-chlorohexan-1-ol
74828
95%
2009-83-8
100.0
1.0300g/mL
98°C
94% min. (GC)
C6H13ClO
Cl(CH2)6OH
100 g
6-chloro-1-hexanol, 6-chlorohexanol, 1-chloro-6-hydroxyhexane, 1-hexanol, 6-chloro, hexamethylene chlorohydrin, 1-chloro-6-hexanol, omega-chlorohexanol, unii-t2hit6n5uf, 6-chloro-hexanol, t2hit6n5uf
C(CCCCl)CCO
136.62
136.62
Liquid

Chemical Identifiers

2009-83-8
136.62
JNTPTNNCGDAGEJ-UHFFFAOYSA-N
74828
C(CCCCl)CCO
C6H13ClO
MFCD00002984
6-chloro-1-hexanol, 6-chlorohexanol, 1-chloro-6-hydroxyhexane, 1-hexanol, 6-chloro, hexamethylene chlorohydrin, 1-chloro-6-hexanol, omega-chlorohexanol, unii-t2hit6n5uf, 6-chloro-hexanol, t2hit6n5uf
6-chlorohexan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-925-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only