missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-5-trimethylsilyl-4-pentyne, 97%

Catalog No. p-7035670
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
AAH5339306 5 g
AAH5339314 25 g
2 options

Catalog No. AAH5339306

Supplier: Thermo Scientific Chemicals H5339306

Only null left

CAS: 77113-48-5 | C8H15ClSi | 174.74 g/mol

1-Chloro-5-trimethylsilyl-4-pentyne is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-5-trimethylsilyl-4-pentyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 77113-48-5
Molecular Formula C8H15ClSi
Molecular Weight (g/mol) 174.74
MDL Number MFCD00671353
InChI Key UPVHIBLXQNNITH-UHFFFAOYSA-N
Synonym 5-chloro-1-pentynyl trimethylsilane, 1-chloro-5-trimethylsilyl-4-pentyne, silane, 5-chloro-1-pentynyl trimethyl, acmc-1bffc, 5-chloro-1-trimethylsilylpent-1-yne, 5-chloropent-1-ynyl trimethyl silane, 5-chloropent-1-yn-1-yl trimethylsilane, silane, 5-chloro-1-pentyn-1-yl trimethyl
PubChem CID 2757942
SMILES C[Si](C)(C)C#CCCCCl

Specifications

Boiling Point 75°C to 77°C (21mmHg)
Quantity 5 g
Linear Formula Cl(CH2)3C≡CSi(CH3)3
Flash Point 65°C (149°F)
IUPAC Name (5-chloropent-1-yn-1-yl)trimethylsilane
Formula Weight 174.75
Percent Purity 97%
Refractive Index 1.456
Density 0.977
Chemical Name or Material 1-Chloro-5-trimethylsilyl-4-pentyne
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only