missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-phenylphthalazine, 96%

Catalog No. AAH3312503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3312503 1 g
AAH3312506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3312503 Supplier Thermo Scientific Chemicals Supplier No. H3312503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10132-01-1 | C14H9ClN2 | 240.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10132-01-1
Molecular Formula C14H9ClN2
Molecular Weight (g/mol) 240.69
MDL Number MFCD00524786
InChI Key WJJDLSHYLZRFDD-UHFFFAOYSA-N
Synonym 1-chloro-4-phenyl-phthalazine, phthalazine,1-chloro-4-phenyl, 4-chloro-1-phenylphthalazine
PubChem CID 272109
IUPAC Name 1-chloro-4-phenylphthalazine
SMILES C1=CC=C(C=C1)C2=NN=C(C3=CC=CC=C32)Cl

Specifications

Melting Point 158°C to 159°C
Quantity 1 g
Formula Weight 240.69
Percent Purity 96%
Chemical Name or Material 1-Chloro-4-phenylphthalazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only