missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-phenylbutane, 97%

CAS: 4830-93-7 | C10H13Cl | 168.664 g/mol

Supplier:  Thermo Scientific Chemicals A1175306

 View more versions of this product

Catalog No. AAA1175306


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Chloro-4-phenylbutane
1.026
104°C (219°F)
1.518
5 g
4-chlorobutyl benzene, 1-chloro-4-phenylbutane, 4-phenylbutyl chloride, 1-4-chlorobutyl benzene, benzene, 4-chlorobutyl, phenylbutylchloride, 1-chloro4-phenylbutane, acmc-209kcr, 4-phenyl-1-chlorobutane, 4-phenyl-n-butyl chloride
C1=CC=C(C=C1)CCCCCl
168.664
168.67
4830-93-7
125°C to 128°C (20 mmHg)
C10H13Cl
MFCD00037117
2042123
FLLZCZIHURYEQP-UHFFFAOYSA-N
4-chlorobutylbenzene
78543
97%

Chemical Identifiers

4830-93-7
168.664
FLLZCZIHURYEQP-UHFFFAOYSA-N
78543
C1=CC=C(C=C1)CCCCCl
C10H13Cl
MFCD00037117
4-chlorobutyl benzene, 1-chloro-4-phenylbutane, 4-phenylbutyl chloride, 1-4-chlorobutyl benzene, benzene, 4-chlorobutyl, phenylbutylchloride, 1-chloro4-phenylbutane, acmc-209kcr, 4-phenyl-1-chlorobutane, 4-phenyl-n-butyl chloride
4-chlorobutylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 225-408-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only