missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-phenylbutane, 97%

Catalog No. AAA1175303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1175303 1 g
AAA1175306 5 g
AAA1175314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1175303 Supplier Thermo Scientific Chemicals Supplier No. A1175303

CAS: 4830-93-7 | C10H13Cl | 168.664 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4830-93-7
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.664
MDL Number MFCD00037117
InChI Key FLLZCZIHURYEQP-UHFFFAOYSA-N
Synonym 4-chlorobutyl benzene, 1-chloro-4-phenylbutane, 4-phenylbutyl chloride, 1-4-chlorobutyl benzene, benzene, 4-chlorobutyl, phenylbutylchloride, 1-chloro4-phenylbutane, acmc-209kcr, 4-phenyl-1-chlorobutane, 4-phenyl-n-butyl chloride
PubChem CID 78543
IUPAC Name 4-chlorobutylbenzene
SMILES C1=CC=C(C=C1)CCCCCl

Specifications

Density 1.026
Boiling Point 125°C to 128°C (20 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.518
Quantity 1 g
Beilstein 2042123
Formula Weight 168.67
Percent Purity 97%
Chemical Name or Material 1-Chloro-4-phenylbutane
EINECSNumber 225-408-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only