missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-iodobutane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. C27135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27135G 5 g
C271325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27135G Supplier TCI America Supplier No. C27135G

Chemical Identifiers

CAS 10297-05-9
Molecular Formula C4H8ClI
Molecular Weight (g/mol) 218.46
MDL Number MFCD00039415
InChI Key JXOSPTBRSOYXGC-UHFFFAOYSA-N
Synonym butane, 1-chloro-4-iodo, tetramethylene chloroiodide, 4-chlorobutyl iodide, 4-iodobutyl chloride, 1-chloro4-iodobutane, 4-chloro-1-iodobutane, 4-iodo-1-chlorobutane, 1-iodo-4-chlorobutane, acmc-1buxg, 1-chloro-4-iodo-butane
PubChem CID 82527
IUPAC Name 1-chloro-4-iodobutane
SMILES ClCCCCI

Specifications

Color Yellow
Boiling Point 67°C
Quantity 5 g
Formula Weight 218.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-4-iodobutane (stabilized with Copper chip)