missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-hydroxyisoquinoline, 97%

Catalog No. AC399780010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399780010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399780010 Supplier Thermo Scientific Chemicals Supplier No. 399780010
Only null left

CAS: 3336-43-4 | C9H6ClNO | 179.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3336-43-4
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.60
MDL Number MFCD03425941
InChI Key JEVLGPVFFYUBRI-UHFFFAOYSA-N
Synonym 1-chloro-4-hydroxyisoquinoline, 4-isoquinolinol, 1-chloro, pubchem18457, 1-chloroisoquinoline-4-ol, 1-chloro-isoquinolin-4-ol, 4-isoquinolinol,1-chloro, 1-chloro-4-hydroxy isoquinoline, 1-chloro-4-hydroxyisoquinoline 97
PubChem CID 4281882
IUPAC Name 1-chloroisoquinolin-4-ol
SMILES OC1=CN=C(Cl)C2=CC=CC=C12

Specifications

Melting Point 195°C to 197°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 179.61
Percent Purity 97%
Chemical Name or Material 1-Chloro-4-hydroxyisoquinoline
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only