missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3-methylbutane 98.0+%, TCI America™
$136.08 - $1118.43
Chemical Identifiers
| CAS | 107-84-6 |
|---|---|
| Molecular Formula | C5H11Cl |
| Molecular Weight (g/mol) | 106.593 |
| MDL Number | MFCD00013693 |
| InChI Key | CZHLPWNZCJEPJB-UHFFFAOYSA-N |
| Synonym | isoamyl chloride, butane, 1-chloro-3-methyl, 3-methylbutyl chloride, isopentyl chloride, 4-chloro-2-methylbutane, 1-chloro-3,3-dimethylpropane, unii-87d3zl9f7a, isoamylchlorid, acmc-2098xt, 1-chloro-3-methyl-butane |
| PubChem CID | 7893 |
| IUPAC Name | 1-chloro-3-methylbutane |
| SMILES | CC(C)CCCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C019925ML
|
TCI America
C019925ML |
25 mL |
Each for $136.08
|
|
|||||
|
C0199500ML
|
TCI America
C0199500ML |
500 mL |
Each for $1,118.43
|
|
|||||
Chemical Identifiers
| 107-84-6 | |
| 106.593 | |
| CZHLPWNZCJEPJB-UHFFFAOYSA-N | |
| 7893 | |
| CC(C)CCCl |
| C5H11Cl | |
| MFCD00013693 | |
| isoamyl chloride, butane, 1-chloro-3-methyl, 3-methylbutyl chloride, isopentyl chloride, 4-chloro-2-methylbutane, 1-chloro-3,3-dimethylpropane, unii-87d3zl9f7a, isoamylchlorid, acmc-2098xt, 1-chloro-3-methyl-butane | |
| 1-chloro-3-methylbutane |
Specifications
| 107-84-6 | |
| 100°C | |
| MFCD00013693 | |
| 1107 | |
| CZHLPWNZCJEPJB-UHFFFAOYSA-N | |
| 1-chloro-3-methylbutane | |
| 7893 | |
| ≥98.0% (GC) | |
| 1-Chloro-3-methylbutane |
| Colorless | |
| C5H11Cl | |
| 25 mL | |
| isoamyl chloride, butane, 1-chloro-3-methyl, 3-methylbutyl chloride, isopentyl chloride, 4-chloro-2-methylbutane, 1-chloro-3,3-dimethylpropane, unii-87d3zl9f7a, isoamylchlorid, acmc-2098xt, 1-chloro-3-methyl-butane | |
| CC(C)CCCl | |
| 106.593 | |
| 106.59 | |
| Liquid |
Safety and Handling
TSCA : Yes