missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3,5-dimethyladamantane 98.0+%, TCI America™

Catalog No. C198125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C198125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C198125G Supplier TCI America Supplier No. C198125G
Only null left

Chemical Identifiers

CAS 707-36-8
Molecular Formula C12H19Cl
Molecular Weight (g/mol) 198.734
InChI Key PXDRFQZLDWZHPX-UHFFFAOYSA-N
PubChem CID 3259679
IUPAC Name 1-chloro-3,5-dimethyladamantane
SMILES CC12CC3CC(C1)(CC(C3)(C2)Cl)C

Specifications

Color Colorless
Boiling Point 87°C
Quantity 25 g
Formula Weight 198.73
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-3,5-dimethyladamantane