missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methylpropane 98.0+%, TCI America™

Catalog No. C020725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C020725ML 25 mL
C0207500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C020725ML Supplier TCI America Supplier No. C020725ML

Chemical Identifiers

CAS 513-36-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000954
InChI Key QTBFPMKWQKYFLR-UHFFFAOYSA-N
PubChem CID 10554
IUPAC Name 1-chloro-2-methylpropane
SMILES CC(C)CCl

Specifications

Color Colorless
Boiling Point 70°C
Quantity 25 mL
UN Number 1127
Synonym isobutyl chloride, propane, 1-chloro-2-methyl, 1-chloro-2-methyl-propane, isobutylchlorid, iso-c4h9cl, sjphadikup@, acmc-1akdy, propane,1-chloro-2-methyl, ksc491k9j, 1-chloro-2-methylpropane
Formula Weight 92.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-methylpropane
EINECSNumber (2)-0060
TSCA Yes