missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methylpropane, 98%

CAS: 513-36-0 | C4H9Cl | 92.566 g/mol

Supplier:  Thermo Scientific Chemicals L0418118

 View more versions of this product

Catalog No. AAL0418118

Only null left
Explore available promotions

Description

Description

1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

1-Chloro-2-methylpropane
-131°C
68°C to 69°C
Sweet
1.3975
50 g
635650
isobutyl chloride, propane, 1-chloro-2-methyl, 1-chloro-2-methyl-propane, isobutylchlorid, iso-c4h9cl, sjphadikup@, acmc-1akdy, propane,1-chloro-2-methyl, ksc491k9j, 1-chloro-2-methylpropane
QTBFPMKWQKYFLR-UHFFFAOYSA-N
1-chloro-2-methylpropane
10554
98%
513-36-0
0.879
−7°C (19°F)
C4H9Cl
MFCD00000954
UN1127
14,5138
Miscible with water,alcohol and diethyl ether.
CC(C)CCl
92.566
92.57

Chemical Identifiers

513-36-0
92.566
QTBFPMKWQKYFLR-UHFFFAOYSA-N
1-chloro-2-methylpropane
C4H9Cl
MFCD00000954
10554
CC(C)CCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES

EINECSNumber : 208-157-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only