missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methylpropane, 98%

Catalog No. AAL0418130
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0418130 250 g
AAL0418118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0418130 Supplier Thermo Scientific Chemicals Supplier No. L0418130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 513-36-0 | C4H9Cl | 92.566 g/mol

1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 513-36-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000954
InChI Key QTBFPMKWQKYFLR-UHFFFAOYSA-N
PubChem CID 10554
IUPAC Name 1-chloro-2-methylpropane
SMILES CC(C)CCl

Specifications

Melting Point -131°C
Density 0.879
Boiling Point 68°C to 69°C
Flash Point −7°C (19°F)
Odor Sweet
Refractive Index 1.3975
Quantity 250 g
UN Number UN1127
Beilstein 635650
Merck Index 14,5138
Synonym isobutyl chloride, propane, 1-chloro-2-methyl, 1-chloro-2-methyl-propane, isobutylchlorid, iso-c4h9cl, sjphadikup@, acmc-1akdy, propane,1-chloro-2-methyl, ksc491k9j, 1-chloro-2-methylpropane
Solubility Information Miscible with water,alcohol and diethyl ether.
Formula Weight 92.57
Percent Purity 98%
Chemical Name or Material 1-Chloro-2-methylpropane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES
EINECSNumber 208-157-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only