missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-2-iodoethane (stabilized with Copper chip) 97.0+%, TCI America™
$221.39 - $1106.97
Chemical Identifiers
| CAS | 624-70-4 |
|---|---|
| Molecular Formula | C2H4ClI |
| Molecular Weight (g/mol) | 190.41 |
| MDL Number | MFCD00041453 |
| InChI Key | JTWWWQGSFTWWDL-UHFFFAOYSA-N |
| Synonym | 1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb |
| PubChem CID | 136438 |
| IUPAC Name | 1-chloro-2-iodoethane |
| SMILES | ClCCI |
Chemical Identifiers
| 624-70-4 | |
| 190.41 | |
| JTWWWQGSFTWWDL-UHFFFAOYSA-N | |
| 136438 | |
| ClCCI |
| C2H4ClI | |
| MFCD00041453 | |
| 1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb | |
| 1-chloro-2-iodoethane |
Specifications
| 624-70-4 | |
| 140°C | |
| MFCD00041453 | |
| 1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb | |
| ClCCI | |
| 190.41 | |
| 190.41 | |
| Liquid |
| Yellow | |
| C2H4ClI | |
| 1 g | |
| JTWWWQGSFTWWDL-UHFFFAOYSA-N | |
| 1-chloro-2-iodoethane | |
| 136438 | |
| ≥97.0% (GC) | |
| 1-Chloro-2-iodoethane (stabilized with Copper chip) |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator