missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodoethane (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. C11901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11901G 1 g
C11905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11901G Supplier TCI America Supplier No. C11901G

Chemical Identifiers

CAS 624-70-4
Molecular Formula C2H4ClI
Molecular Weight (g/mol) 190.41
MDL Number MFCD00041453
InChI Key JTWWWQGSFTWWDL-UHFFFAOYSA-N
Synonym 1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
PubChem CID 136438
IUPAC Name 1-chloro-2-iodoethane
SMILES ClCCI

Specifications

Color Yellow
Boiling Point 140°C
Quantity 1 g
Formula Weight 190.41
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-iodoethane (stabilized with Copper chip)
TSCA No
Recommended Storage Refrigerator