missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodoethane, 96%, stab. with copper

Catalog No. AAL1074203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1074203 1 g
AAL1074206 5 g
AAL1074214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1074203 Supplier Thermo Scientific Chemicals Supplier No. L1074203

CAS: 624-70-4 | C2H4ClI | 190.41 g/mol

1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

Solubility
Soluble inorganic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 624-70-4
Molecular Formula C2H4ClI
Molecular Weight (g/mol) 190.41
MDL Number MFCD00041453
InChI Key JTWWWQGSFTWWDL-UHFFFAOYSA-N
Synonym 1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
PubChem CID 136438
IUPAC Name 1-chloro-2-iodoethane
SMILES ClCCI

Specifications

Density 2.088
Boiling Point 61°C to 62°C (30 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.566
Quantity 1 g
Beilstein 1730865
Sensitivity Light sensitive
Solubility Information Soluble inorganic solvents. Insoluble in water.
Formula Weight 190.41
Percent Purity 96%
Chemical Name or Material 1-Chloro-2-iodoethane
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 677-827-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only