missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-ethynylbenzene 98.0+%, TCI America™

Catalog No. p-7125630
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27501G 1 g
C27505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27501G Supplier TCI America Supplier No. C27501G
Only null left

Chemical Identifiers

CAS 873-31-4
Molecular Formula C8H5Cl
Molecular Weight (g/mol) 136.578
MDL Number MFCD00269951
InChI Key DGLHLIWXYSGYBI-UHFFFAOYSA-N
Synonym 2-chlorophenylacetylene, 2'-chlorophenyl acetylene, 1-chloro-2-ethynyl-benzene, benzene, 1-chloro-2-ethynyl, 1-chlor-2-ethinylbenzol, o-chlorphenylacetylen, o-chlorophenylacetylene, pubchem2550, 2-chlorophenyl acetylene, 2;-chlorophenylacetylene
PubChem CID 585995
IUPAC Name 1-chloro-2-ethynylbenzene
SMILES C#CC1=CC=CC=C1Cl

Specifications

Quantity 1 g
Physical Form Clear Liquid at 20°C
Boiling Point 72°C
Formula Weight 136.58
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1-Chloro-2-ethynylbenzene
TSCA No
Recommended Storage Refrigerator