missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-2,2,2-trifluoroethyl Difluoromethyl Ether 98.0+%, TCI America™
$86.64 - $86.64
Chemical Identifiers
| CAS | 26675-46-7 |
|---|---|
| Molecular Formula | C3H2ClF5O |
| Molecular Weight (g/mol) | 184.49 |
| MDL Number | MFCD00066609 |
| InChI Key | PIWKPBJCKXDKJR-UHFFFAOYNA-N |
| Synonym | isoflurane, forane, 1-chloro-2,2,2-trifluoroethyl difluoromethyl ether, forene, aerrane, isoflurano, isofluranum, isoflo, compound 469, 2-chloro-2-difluoromethoxy-1,1,1-trifluoroethane |
| PubChem CID | 3763 |
| ChEBI | CHEBI:6015 |
| IUPAC Name | 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane |
| SMILES | FC(F)OC(Cl)C(F)(F)F |
Chemical Identifiers
| 26675-46-7 | |
| 184.49 | |
| PIWKPBJCKXDKJR-UHFFFAOYNA-N | |
| 3763 | |
| 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane |
| C3H2ClF5O | |
| MFCD00066609 | |
| isoflurane, forane, 1-chloro-2,2,2-trifluoroethyl difluoromethyl ether, forene, aerrane, isoflurano, isofluranum, isoflo, compound 469, 2-chloro-2-difluoromethoxy-1,1,1-trifluoroethane | |
| CHEBI:6015 | |
| FC(F)OC(Cl)C(F)(F)F |
Specifications
| 26675-46-7 | |
| 49°C | |
| MFCD00066609 | |
| isoflurane, forane, 1-chloro-2,2,2-trifluoroethyl difluoromethyl ether, forene, aerrane, isoflurano, isofluranum, isoflo, compound 469, 2-chloro-2-difluoromethoxy-1,1,1-trifluoroethane | |
| FC(F)OC(Cl)C(F)(F)F | |
| 184.49 | |
| CHEBI:6015 | |
| ≥98.0% (GC) | |
| 1-Chloro-2,2,2-trifluoroethyl Difluoromethyl Ether |
| Colorless | |
| C3H2ClF5O | |
| 5 g | |
| PIWKPBJCKXDKJR-UHFFFAOYNA-N | |
| 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane | |
| 3763 | |
| 184.49 | |
| Liquid |
Safety and Handling
RTECSNumber : KN6799000
TSCA : No