Learn More
1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, 97%
CAS: 26675-46-7 | C3H2ClF5O | 184.49 g/mol
$340.70 - $340.70
Chemical Identifiers
| CAS | 26675-46-7 |
|---|---|
| Molecular Formula | C3H2ClF5O |
| Molecular Weight (g/mol) | 184.49 |
| MDL Number | MFCD00066609 |
| InChI Key | PIWKPBJCKXDKJR-UHFFFAOYNA-N |
| Synonym | isoflurane, forane, 1-chloro-2,2,2-trifluoroethyl difluoromethyl ether, forene, aerrane, isoflurano, isofluranum, isoflo, compound 469, 2-chloro-2-difluoromethoxy-1,1,1-trifluoroethane |
| PubChem CID | 3763 |
| ChEBI | CHEBI:6015 |
| IUPAC Name | 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane |
| SMILES | FC(F)OC(Cl)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1731514
|
Thermo Scientific Chemicals
L1731514 |
25 g |
Each for $340.70
|
|
|||||
Description
1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether is a useful solvent for dispersing fluorinated materials.
For Research and Development use only. Not to be used in clinical studies
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 26675-46-7 | |
| 184.49 | |
| PIWKPBJCKXDKJR-UHFFFAOYNA-N | |
| 3763 | |
| 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane |
| C3H2ClF5O | |
| MFCD00066609 | |
| isoflurane, forane, 1-chloro-2,2,2-trifluoroethyl difluoromethyl ether, forene, aerrane, isoflurano, isofluranum, isoflo, compound 469, 2-chloro-2-difluoromethoxy-1,1,1-trifluoroethane | |
| CHEBI:6015 | |
| FC(F)OC(Cl)C(F)(F)F |
Specifications
| 26675-46-7 | |
| 1.496g/mL | |
| Ether-like | |
| <1.3 | |
| 25 g | |
| 14,5175 | |
| PIWKPBJCKXDKJR-UHFFFAOYNA-N | |
| 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane | |
| 3763 | |
| 184.49 | |
| 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether |
| Colorless | |
| 48°C to 49°C | |
| C3H2ClF5O | |
| MFCD00066609 | |
| 1852087 | |
| isoflurane, forane, 1-chloro-2,2,2-trifluoroethyl difluoromethyl ether, forene, aerrane, isoflurano, isofluranum, isoflo, compound 469, 2-chloro-2-difluoromethoxy-1,1,1-trifluoroethane | |
| FC(F)OC(Cl)C(F)(F)F | |
| 184.49 | |
| CHEBI:6015 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335-H336
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : KN6799000
RUO – Research Use Only