missing translation for 'onlineSavingsMsg'
Learn More

1-Carbobenzoxypiperazine 95.0+%, TCI America™

Catalog No. C21785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21785G 5 g
C217825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21785G Supplier TCI America Supplier No. C21785G

Chemical Identifiers

CAS 31166-44-6
Molecular Formula C12H17N2O2
Molecular Weight (g/mol) 221.28
MDL Number MFCD00274317
InChI Key CTOUWUYDDUSBQE-UHFFFAOYSA-O
Synonym 1-cbz-piperazine, benzyl 1-piperazinecarboxylate, 1-benzyloxycarbonyl piperazine, 1-carbobenzoxypiperazine, n-cbz-piperazine, 1-z-piperazine, 1-piperazinecarboxylic acid, phenylmethyl ester, piperazine-1-carboxylic acid benzyl ester, phenylmethyl piperazinecarboxylate, cbz-piperazine
PubChem CID 643495
IUPAC Name 4-[(benzyloxy)carbonyl]piperazin-1-ium
SMILES O=C(OCC1=CC=CC=C1)N1CC[NH2+]CC1

Specifications

Boiling Point 161°C
Quantity 5 g
Formula Weight 220.27
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Carbobenzoxypiperazine