missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Carbobenzoxy-1,2,3,6-tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
$161.72 - $483.21
Chemical Identifiers
| CAS | 286961-15-7 |
|---|---|
| Molecular Formula | C19H26BNO4 |
| Molecular Weight (g/mol) | 343.23 |
| MDL Number | MFCD11521562 |
| InChI Key | QDSFHRPYZPQWEJ-UHFFFAOYSA-N |
| Synonym | benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylic acid benzyl ester, n-cbz-3,6-dihydro-2h-pyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, 1-benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydropyridine-1 2h-carboxylate, benzyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, n-cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, benzyl 4-4,4,5,5-tetrameth |
| PubChem CID | 11290836 |
| IUPAC Name | benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OCC3=CC=CC=C3 |
Chemical Identifiers
| 286961-15-7 | |
| 343.23 | |
| QDSFHRPYZPQWEJ-UHFFFAOYSA-N | |
| 11290836 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OCC3=CC=CC=C3 |
| C19H26BNO4 | |
| MFCD11521562 | |
| benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylic acid benzyl ester, n-cbz-3,6-dihydro-2h-pyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, 1-benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydropyridine-1 2h-carboxylate, benzyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, n-cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, benzyl 4-4,4,5,5-tetrameth | |
| benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate |
Specifications
| 286961-15-7 | |
| White-Yellow | |
| MFCD11521562 | |
| benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylic acid benzyl ester, n-cbz-3,6-dihydro-2h-pyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, 1-benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydropyridine-1 2h-carboxylate, benzyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, n-cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, benzyl 4-4,4,5,5-tetrameth | |
| B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OCC3=CC=CC=C3 | |
| 343.23 | |
| 343.23 | |
| Crystalline Powder |
| 95°C | |
| C19H26BNO4 | |
| 1 g | |
| QDSFHRPYZPQWEJ-UHFFFAOYSA-N | |
| benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate | |
| 11290836 | |
| ≥98.0% (GC,T) | |
| 1-Carbobenzoxy-1,2,3,6-tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Safety and Handling
TSCA : No