missing translation for 'onlineSavingsMsg'
Learn More

1-Carbobenzoxy-1,2,3,6-tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. C32435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32435G 5 g
C32431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32435G Supplier TCI America Supplier No. C32435G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 286961-15-7
Molecular Formula C19H26BNO4
Molecular Weight (g/mol) 343.23
MDL Number MFCD11521562
InChI Key QDSFHRPYZPQWEJ-UHFFFAOYSA-N
Synonym benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylic acid benzyl ester, n-cbz-3,6-dihydro-2h-pyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, 1-benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydropyridine-1 2h-carboxylate, benzyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, n-cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, benzyl 4-4,4,5,5-tetrameth
PubChem CID 11290836
IUPAC Name benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OCC3=CC=CC=C3

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 5 g
Formula Weight 343.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Carbobenzoxy-1,2,3,6-tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine