missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butylpyridinium Tetrafluoroborate 94.0+%, TCI America™
$216.24 - $741.89
Chemical Identifiers
| CAS | 203389-28-0 |
|---|---|
| Molecular Formula | C9H14BF4N |
| Molecular Weight (g/mol) | 223.022 |
| MDL Number | MFCD03427602 |
| InChI Key | XLWDQAHXRCBPEE-UHFFFAOYSA-N |
| Synonym | 1-butylpyridinium tetrafluoroborate, 1-butylpyridin-1-ium tetrafluoroborate, n-butylpyridinium tetrafluoroborate, bpmbf4, acmc-1co7o, dsstox_cid_29097, dsstox_rid_83316, dsstox_gsid_49241, ksc199a6h, 1-butylpyridin-1-ium; tetrafluoroboranuide |
| PubChem CID | 2734180 |
| IUPAC Name | 1-butylpyridin-1-ium;tetrafluoroborate |
| SMILES | [B-](F)(F)(F)F.CCCC[N+]1=CC=CC=C1 |
Chemical Identifiers
| 203389-28-0 | |
| 223.022 | |
| XLWDQAHXRCBPEE-UHFFFAOYSA-N | |
| 2734180 | |
| [B-](F)(F)(F)F.CCCC[N+]1=CC=CC=C1 |
| C9H14BF4N | |
| MFCD03427602 | |
| 1-butylpyridinium tetrafluoroborate, 1-butylpyridin-1-ium tetrafluoroborate, n-butylpyridinium tetrafluoroborate, bpmbf4, acmc-1co7o, dsstox_cid_29097, dsstox_rid_83316, dsstox_gsid_49241, ksc199a6h, 1-butylpyridin-1-ium; tetrafluoroboranuide | |
| 1-butylpyridin-1-ium;tetrafluoroborate |
Specifications
| 203389-28-0 | |
| C9H14BF4N | |
| 5 g | |
| 1-butylpyridinium tetrafluoroborate, 1-butylpyridin-1-ium tetrafluoroborate, n-butylpyridinium tetrafluoroborate, bpmbf4, acmc-1co7o, dsstox_cid_29097, dsstox_rid_83316, dsstox_gsid_49241, ksc199a6h, 1-butylpyridin-1-ium; tetrafluoroboranuide | |
| [B-](F)(F)(F)F.CCCC[N+]1=CC=CC=C1 | |
| 223.022 | |
| 223.02 | |
| Liquid |
| Yellow | |
| MFCD03427602 | |
| 1760 | |
| XLWDQAHXRCBPEE-UHFFFAOYSA-N | |
| 1-butylpyridin-1-ium;tetrafluoroborate | |
| 2734180 | |
| ≥94.0% (N) | |
| 1-Butylpyridinium Tetrafluoroborate |
Safety and Handling
TSCA : No