missing translation for 'onlineSavingsMsg'
Learn More

1-Butylpyridinium Chloride 98.0+%, TCI America™

Catalog No. B132910G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B132910G 10 g
B132925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B132910G Supplier TCI America Supplier No. B132910G

Chemical Identifiers

CAS 1124-64-7
Molecular Formula C9H14ClN
Molecular Weight (g/mol) 171.668
MDL Number MFCD00060047
InChI Key POKOASTYJWUQJG-UHFFFAOYSA-M
Synonym 1-butylpyridinium chloride, 1-butylpyridin-1-ium chloride, n-butylpyridinium chloride, butylpyridinium chloride, dsstox_cid_11462, dsstox_rid_78879, dsstox_gsid_31462, unii-f8m63j351a, butylpyridiniumchloride, butylpyridine, chloride
PubChem CID 2734171
IUPAC Name 1-butylpyridin-1-ium;chloride
SMILES CCCC[N+]1=CC=CC=C1.[Cl-]

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 10 g
Formula Weight 171.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butylpyridinium Chloride