missing translation for 'onlineSavingsMsg'
Learn More

1-Butylpiperazine 98.0+%, TCI America™

Catalog No. B40785ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40785ML 5 mL
B407825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40785ML Supplier TCI America Supplier No. B40785ML
Only null left

Chemical Identifiers

CAS 5610-49-1
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00039702
InChI Key YKSVXVKIYYQWBB-UHFFFAOYSA-N
PubChem CID 424322
IUPAC Name 1-butylpiperazine
SMILES CCCCN1CCNCC1

Specifications

Color Yellow
Boiling Point 192°C
Quantity 5 mL
Formula Weight 142.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Butylpiperazine