missing translation for 'onlineSavingsMsg'
Learn More

1-Butylphosphonic acid, 98%

Catalog No. AAA1798303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1798303 1 g
AAA1798306 5 g
AAA1798314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1798303 Supplier Thermo Scientific Chemicals Supplier No. A1798303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3321-64-0 | C4H11O3P | 138.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3321-64-0
Molecular Formula C4H11O3P
Molecular Weight (g/mol) 138.10
MDL Number MFCD00012295
InChI Key UOKRBSXOBUKDGE-UHFFFAOYSA-N
Synonym n-butylphosphonic acid, 1-butanephosphonic acid, butylphosphonate, 1-butylphosphonic acid, phosphonic acid, butyl, n-butyl phosphonic acid, acmc-1cr41
PubChem CID 76839
ChEBI CHEBI:41384
IUPAC Name butylphosphonic acid
SMILES CCCCP(O)(O)=O

Specifications

Melting Point 105°C to 107°C
Odor Odorless
Quantity 1 g
UN Number UN3261
Beilstein 1749026
Formula Weight 138.1
Percent Purity 98%
Chemical Name or Material 1-Butylphosphonic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only