missing translation for 'onlineSavingsMsg'
Learn More

1-Butylimidazole 98.0+%, TCI America™

Catalog No. B234525G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B234525G 25 g
B2345100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B234525G Supplier TCI America Supplier No. B234525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4316-42-1
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00042753
InChI Key MCMFEZDRQOJKMN-UHFFFAOYSA-N
Synonym 1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
PubChem CID 61347
IUPAC Name 1-butyl-1H-imidazole
SMILES CCCCN1C=CN=C1

Specifications

Color Yellow
Boiling Point 235°C
Quantity 25 g
UN Number 2690
Formula Weight 124.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Butylimidazole